Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Colistin sulfate, 50mg/mL in distilled water, sterile-filtered

Click to view available options
Quantity:
1mL
5 x 1mL
1264-72-8
MFCD00146495
1264-72-8
MFCD00146495
Physical Form Liquid
Quantity 5 x 1mL
Chemical Name or Material Colistin sulfate, 50mg/mL in distilled water, sterile-filtered

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : TR1500000

storageNote1 : -30° to -10°C