Colitag™ Presence/Absence (P/A) Water Test Kit

Definitive water test system for coliforms and E. coli

Manufacturer: CPI INTERNATIONAL INC 46000013

 View more versions of this product

Catalog No. 46000013

  Check Availability
Add to Cart

    Specifications

    Shelf Life 1 Year
    Method ONPG/MUG
    Product Type Colitag P/A Test Kit
    For Use With (Application) For coliforms, E. coli
    No. of Tests 100.0
    Storage Requirements 4° to 7°C
    Certifications/Compliance EPA

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit