Learn More
Abnova™ COLL2-1 (Human) ELISA Kit
Competitive enzyme immunoassay for the quantitative measurement of a degradation fragment containing a 9 amino acid sequence (HRGYPGLDG) from type II collagen molecule
Manufacturer:  Abnova™ KA4900
Catalog No. 89-032-265
Add to cart
Description
Description
  • For the quantification of Coll2-1, a degradation product of type II collagen in human serum
Specifications
Specifications
COLL2-1 (Human) ELISA Kit is a competitive enzyme immunoassay for the quantitative measurement of a degradation fragment containing a 9 amino acid sequence (HRGYPGLDG) from type II collagen molecule.
Quantification
Store at 4°C