Learn More
Collagen Hydrolysate, Powder, Spectrum™
C1953, 46022-05-3
Manufacturer:  Spectrum Chemical Manufacturing CorporationSupplier Diversity Partner C1953500GM
 View more versions of this product
Catalog No. 18-601-812
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Spectrum™ Collagen Hydrolysate, Powder, also known as hydrolyzed collagen, is produced from collagen found in bones. In cosmetics, collagen hydrolysate is used in topical creams, acting as a product texture conditioner, and moisturizer. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications
Specifications
0.98
500 g