Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gibco™ Collagen I, bovine
Collagen I Bovine Protein
Supplier: Gibco™ A1064401
Description
Collagen I, Bovine is a naturally occurring extra cellular matrix found in tendons and other connective tissues. It plays a key role in cell growth, differentiation, attachment, and migration. This solution is supplied at 5 mg⁄ml in a 10 ml vial providing the flexibility to dilute to lower concentrations. The detailed gelling procedure included in the package insert allows the user to form a firm, clear gel for 3D applications.Specifications
5 mg/mL | |
10 mL | |
Bovine | |
≤10 EU/mL, ≤ 10 EU/ml | |
Native | |
Collagen | |
Gibco™ |
Cell Culture | |
Collagen | |
Store in refrigerator (2–8°C) and protect from light. | |
Wet Ice | |
Liquid | |
Extracellular Matrix & Cell Adhesion |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?
Provide Content Correction