Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Collagen II Monoclonal Antibody (5B2.5), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA512781
Description
MA5-12781 targets Chicken Collagen II in IF and WB applications and shows reactivity with Chicken, Human, and mouse samples. The MA5-12781 immunogen is a purified preparation of lathyritic type II collagen from embryonic chicken sternum.
Collagen II Monoclonal antibody specifically detects Collagen II in Chicken, Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.
Specifications
Collagen II | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P02458, P02460, P28481 | |
Col2a1 | |
A purified preparation of lathyritic type II collagen from embryonic chicken sternum | |
500 μL | |
Primary | |
Chicken, Human, Mouse | |
Antibody | |
IgG2a κ |
Immunocytochemistry, Western Blot | |
5B2.5 | |
Unconjugated | |
Col2a1 | |
alpha 1 type II procollagen; alpha 1 type IIA collagen; alpha 1 type IIA collagen precursor; alpha collagen type II; alpha-1 type II collagen; ANFH; AOM; arthroophthalmopathy, progressive (Stickler syndrome); cartilage collagen; CB11; CG2A1A; CGA; Chondrocalcin; COL11A3; Col2; Col2a; col2a1; Col2a-1; col2a1.L; col2a1a; col2a1-a; col2a1-b; Collagen 2; Collagen alpha-1(II) chain; collagen II, alpha-1 polypeptide; collagen type II (Col2A1) gene, enhancer region; collagen type II alpha 1 chain; collagen type IIA; collagen, type II, alpha 1; collagen, type II, alpha 1 (primary osteoarthritis, spondyloepiphyseal dysplasia, congenital); collagen, type II, alpha 1 L homeolog; COLLII; cyanogen bromide; Del1; disproportionate micromelia; Dmm; Lpk; M100856; MGC131516; pro alpha1 type II collagen; pro-alpha 1(II); pro-alpha-1 type II collagen; procollagen alpha 1(II); Procollagen II alpha 1; procollagen, type II, alpha 1; Rgsc856; SEDC; STL1; type II collagen; type II collagen cyanogen bromide fragment CB10; type II collogen cyanogen bromide fragment CB8; unnamed protein product; X-col II; xcol2a1; XELAEV_18013523mg | |
Mouse | |
Protein A | |
RUO | |
1280, 12824, 395069 | |
4° C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction