Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Collagen IV Ab-3, Mouse Monoclonal Antibody

Obtain accurate and reproducible results in immunohistochemistry applications with Thermo Scientific™ Collagen IV Ab-3, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ MS747R7

 View more versions of this product

Catalog No. MS747R7

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Obtain accurate and reproducible results in immunohistochemistry applications with Thermo Scientific™ Collagen IV Ab-3, Mouse Monoclonal Antibody, one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Collagen IV is a major constituent of the basement membranes along with laminins and enactins. It is composed of α1(IV) chain and α2(IV) chain in 2:1 ratio . It can form insoluble fibers with high tensile strength. Antibody to collagen IV is useful in detecting the loss of parts of basement membrane in carcinomas.

Host Species: Mouse

Clone: PHM-12 + CIV 22

Isotype: IgG1 + IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Human glomeruli

Positive Control: Skin

Cellular Localization: Basement membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Collagen IV Ab-3
Monoclonal
Unconjugated
Murine
IgG1
IVD
Primary
Monoclonal
Immunohistochemistry (Paraffin)
PHM-12+CIV 22
Prediluted
Human glomeruli
7mL
Extracellular Matrix
Primary
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit