Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen IV alpha 1, Mouse anti-Human, Biotin, Clone: 577222, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals MAB63081B

Catalog No. NB003190


Add to cart

Description

Description

Collagen IV alpha 1 Monoclonal specifically detects Collagen IV alpha 1 in Human samples. It is validated for Immunocytochemistry.
Specifications

Specifications

Collagen IV alpha 1
577222
Immunocytochemistry
Purified, Conjugated
COL4A1
Partial recombinant human Collagen IV alpha 1 protein (amino acids 1441-1669) [UniProt P02462].
Protein A or G purified
RUO
Store at 4°C in the dark.
Monoclonal
Immunocytochemistry
Biotin
PBS with 0.05% Sodium Azide
alpha-1 chain, arresten, COL4A1 NC1 domain, collagen alpha-1(IV) chain, collagen IV, alpha-1 polypeptide, collagen, type IV, alpha 1, Collagen-4, EC 3.4.23, EC 6.3.1.2
Mouse
IgG1
0.1 mL
Cell Biology, Cellular Markers, Extracellular Matrix, Stem Cell Markers
Primary
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit