Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen IV alpha 1 Mouse anti-Human, Clone: IHC549, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP2759827ML

 View more versions of this product

Catalog No. NBP2759827M

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Collagen IV alpha 1 Monoclonal antibody specifically detects Collagen IV alpha 1 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications

Specifications

Collagen IV alpha 1
IHC549
Immunohistochemistry 1:100 - 1:200, Immunohistochemistry-Paraffin 1:100 - 1:200
alpha-1 chain, arresten, COL4A1 NC1 domain, collagen alpha-1(IV) chain, collagen IV, alpha-1 polypeptide, collagen, type IV, alpha 1, Collagen-4, EC 3.4.23, EC 6.3.1.2
Mouse
IgG1 Kappa
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Tris Buffer, pH 7.3 - 7.7, with 1% BSA with 0.1% sodium azide
COL4A1
Human Placenta
7 mL
Primary
1282
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit