Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Collagen IV Antibody (IHC549), Novus Biologicals™
Mouse Monoclonal Antibody
$229.00
Specifications
Antigen | Collagen IV alpha 1 |
---|---|
Clone | IHC549 |
Dilution | Immunohistochemistry 1:100 - 1:200, Immunohistochemistry-Paraffin 1:100 - 1:200 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
N27598201M
|
Novus Biologicals
NBP2759820.1ML |
0.1 mL |
Each of 1 for $229.00
|
|
|||||
Description
Collagen IV Monoclonal specifically detects Collagen IV in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Collagen IV alpha 1 | |
Immunohistochemistry 1:100 - 1:200, Immunohistochemistry-Paraffin 1:100 - 1:200 | |
Monoclonal | |
Mouse | |
Tris Buffer, pH 7.3 - 7.7, with 1% BSA with 0.1% sodium azide | |
1282 | |
Human Placenta | |
Primary |
IHC549 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Human | |
alpha-1 chain, arresten, COL4A1 NC1 domain, collagen alpha-1(IV) chain, collagen IV, alpha-1 polypeptide, collagen, type IV, alpha 1, Collagen-4, EC 3.4.23, EC 6.3.1.2 | |
COL4A1 | |
IgG1 κ | |
Store at 4°C in the dark. Do not freeze. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title