Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen IV alpha 1 Mouse anti-Human, PE-Cyanine7, Clone: 1043, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP197716PECY7

Catalog No. N197716PEC7

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Collagen IV alpha 1 Monoclonal antibody specifically detects Collagen IV alpha 1 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Collagen IV alpha 1
1043
Flow Cytometry
Conjugated, Purified
COL4A1
The immunogen has been isolated from human placenta. Collagen Type IV recognizes the extracellular - basemembranes.
Protein A or G purified
Cytoskeleton Markers, Extracellular Matrix, Stem Cell Markers
Primary
1282.0
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
alpha-1 chain, arresten, COL4A1 NC1 domain, collagen alpha-1(IV) chain, collagen IV, alpha-1 polypeptide, collagen, type IV, alpha 1, Collagen-4, EC 3.4.23, EC 6.3.1.2
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit