Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Collagen IV Monoclonal Antibody (CIV 22), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA513437
Description
MA5-13437 targets Collagen IV in IHC (P) applications and shows reactivity with Bovine and Human samples. The MA5-13437 immunogen is human glomeruli.
Collagen IV Monoclonal antibody specifically detects Collagen IV in Bovine, Human samples. It is validated for Immunohistochemistry (Paraffin).
Specifications
Collagen IV | |
Monoclonal | |
Conc. Not Determined | |
tissue culture supernatant with 15mM sodium azide | |
P02462, P08572, P29400, P53420, Q01955, Q28084, Q29442, Q7SIB2, Q7SIB3 | |
COL4A1, COL4A2, Col4a3, COL4A4, COL4A5 | |
Human glomeruli | |
RUO | |
1282, 1284, 1285, 1286, 1287, 282191, 317711, 407107, 508632, 511602 | |
4° C | |
Liquid |
Immunohistochemistry (Paraffin) | |
CIV 22 | |
Unconjugated | |
COL4A2 | |
[a]3(IV); [a]4(IV); alpha 1 type IV collagen; alpha type IV collagen basement membrane; alpha1(IV) collagen; alpha-2 collagen type IV; alpha-3 type IV collagen; alpha3(IV); Arresten; Bru; BSVD; CA44; Canstatin; Col4a1; Col4a-1; COL4A1 NC1 domain; Col4a2; Col4a-2; COL4A3; col4a4; Collagen 4; Collagen alpha-1(IV) chain; collagen alpha-2(IV) chain; Collagen alpha-3(IV) chain; collagen alpha-4(IV) chain; collagen IV, alpha-1 polypeptide; collagen IV, alpha-3 polypeptide; Collagen IV, alpha-4 polypeptide; collagen of basement membrane, alpha-1 chain; collagen of basement membrane, alpha-4 chain; collagen type IV alpha 1 chain; collagen type IV alpha 2; collagen type IV alpha 2 chain; collagen type IV alpha 3 chain; collagen type IV alpha 4 chain; collagen type IV alpha3 chain; collagen, type IV, alpha 1; collagen, type IV, alpha 2; collagen, type IV, alpha 3; collagen, type IV, alpha 3 (Goodpasture antigen); collagen, type IV, alpha 4; Del(8)44H; Del(8)Bru44H; E130010M05Rik; Goodpasture antigen; ICH; LOW QUALITY PROTEIN: collagen alpha-1(IV) chain; LOW QUALITY PROTEIN: collagen alpha-3(IV) chain; LOW QUALITY PROTEIN: collagen alpha-4(IV) chain; POREN2; procollagen alpha1(IV); procollagen, type IV, alpha 1; procollagen, type IV, alpha 2; procollagen, type IV, alpha 3; procollagen, type IV, alpha 4; RATOR; Raw; retinal anterior wiring; Svc; tumstatin; Type IV Collagen | |
Mouse | |
500 μL | |
Primary | |
Bovine, Human | |
Antibody | |
IgG1 |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction