Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen IV Mouse anti-Human, Clone: 1042, eBioscience™ Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  LIFE TECHNOLOGIES 14987195

Encompass

 View more versions of this product

Catalog No. 14-987-195


Add to cart

Description

Description

This monoclonal antibody 1042 recognizes human collagen Type IV. Collagens are the main components of the extracellular matrix and provide the tensile strength to tissues. There are four main types of collagen (I, II, III and IV) which contribute to the connective tissue. Expression of type IV is restricted to the basal lamina. It contains two alpha-1 and one alpha-2-like chains. Collagen Type IV is associated with disorders such as Alport syndrome and Goodpasture's syndrome. Applications Reported: This 1042 antibody has been reported for use in immunoprecipitation, western blotting, immunohistochemical staining of frozen (IHC-F) and formalin-fixed paraffin embedded (IHC-P) tissue sections. Applications Tested: This 1042 antibody has been tested by immunohistochemistry on formalin-fixed paraffin embedded (FFPE) tissue with low pH antigen retrieval. This can be used at less than or equal to 5 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 µm post-manufacturing filtered.

Collagen IV Monoclonal antibody specifically detects Collagen IV in Human samples. It is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Specifications

Specifications

Collagen IV
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P02462, P08572, P53420, Q01955
Mouse
10 mg
Primary
Human
Antibody
IgG2b κ
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
1042
Unconjugated
COL4A4
[a]3(IV); [a]4(IV); alpha 1 type IV collagen; alpha type IV collagen basement membrane; alpha1(IV) collagen; alpha-2 collagen type IV; alpha-3 type IV collagen; alpha3(IV); Arresten; Bru; BSVD; CA44; Canstatin; COL4A1; Col4a-1; COL4A1 NC1 domain; Col4a2; Col4a-2; COL4A3; col4a4; Collagen 4; Collagen alpha-1(IV) chain; collagen alpha-2(IV) chain; Collagen alpha-3(IV) chain; collagen alpha-4(IV) chain; collagen IV, alpha-1 polypeptide; collagen IV, alpha-3 polypeptide; Collagen IV, alpha-4 polypeptide; collagen of basement membrane, alpha-1 chain; collagen of basement membrane, alpha-4 chain; collagen type IV alpha 1 chain; collagen type IV alpha 2; collagen type IV alpha 3 chain; collagen type IV alpha 4 chain; collagen type IV alpha3 chain; collagen, type IV, alpha 1; collagen, type IV, alpha 2; collagen, type IV, alpha 3; collagen, type IV, alpha 3 (Goodpasture antigen); collagen, type IV, alpha 4; Del(8)44H; Del(8)Bru44H; E130010M05Rik; Goodpasture antigen; ICH; POREN2; procollagen alpha1(IV); procollagen, type IV, alpha 1; procollagen, type IV, alpha 2; procollagen, type IV, alpha 3; procollagen, type IV, alpha 4; RATOR; Raw; retinal anterior wiring; Svc; Tumstatin; Type IV Collagen
Affinity chromatography
RUO
1282, 1284, 1285, 1286
4° C
Liquid
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit