Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen Type I Rabbit anti-Porcine, Polyclonal, Biologo Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

$558.76 - $1064.80

Specifications

Antigen Collagen Type I
Concentration 1 mg/mL
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Radioimmune Assays, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-210-0717 Non-distribution product as customer accommodation.
View Documents
Exalpha Biologicals IncSupplier Diversity Partner
CO20191
0.5 mL
Each for $1,064.80
Add to cart
 
50-210-0718 Non-distribution product as customer accommodation.
View Documents
Exalpha Biologicals IncSupplier Diversity Partner
CO2019101
0.1 mL
Each for $558.76
Add to cart
 
Description

Description

Collagen Type I Polyclonal antibody specifically detects Collagen Type I in Porcine samples. It is validated for ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Radioimmune Assays, Western Blot
Specifications

Specifications

Collagen Type I
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Radioimmune Assays, Western Blot
Unconjugated
Rabbit
Porcine
AA960264; AI325291; alpha 1 type I collagen; alpha 2 type I procollagen; alpha 2(I) procollagen; alpha 2(I)-collagen; alpha1 chain of type I collagen; alpha-1 type 1 collagen; alpha-1 type I collagen; alpha1(I) procollagen; alpha2 chain of type I collagen; Alpha-2 type I collagen; alpha2(I) collagen; COL1; COL1A1; Col1a-1; COL1A1; matrix protein; Col1a2; Col1a-2; Cola1; Cola-1; Cola2; Cola-2; COLIA1; collagen 1 alpha 2 chain; collagen alpha 1 chain type I; collagen alpha-1(I) chain; collagen alpha-1(I) chain preproprotein; Collagen alpha-2(I) chain; collagen COL1A2; collagen I alpha-2 polypeptide; collagen I, alpha-2 polypeptide; collagen of skin, tendon and bone, alpha-1 chain; collagen of skin, tendon and bone, alpha-2 chain; collagen type I alpha 1; collagen type I alpha 1 chain; collagen type I alpha 2 chain; collagen, type 1, alpha 1; collagen, type I, alpha 1; collagen, type I, alpha 2; EDSC; LOC100347598; Mov13; Mov-13; OI1; OI2; OI3; OI4; oim; osteogenesis imperfecta; pro-alpha-1 collagen type 1; procollagen type I, alpha 1; procollagen, type 1, alpha 1; procollagen, type I, alpha 1; procollagen, type I, alpha 2; similar to pro alpha 1(I) collagen; Type I Collagen; type I collagen alpha1; type I collagen alpha-2 chain; type I proalpha 1; type I procollagen; type I procollagen alpha 1 chain; type I-alpha 1 collagen
Purified collagen type I from porcine tendon
Polyclonal
Affinity chromatography
Antibody
1 mg/mL
Polyclonal
Lyophilized
RUO
PBS with no preservative; pH
100626716, 100738123
IgG
Primary
-20°C
COL1A2
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit