Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen Type II Mouse Anti-Human, Unlabeled, Clone: II-4C11, MP Biologicals

Mouse Monoclonal Antibody

$284.50 - $708.50

Specifications

Antigen Collagen Type II
Clone II-4C11
Concentration 1.0mg/mL
Format Purified
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ICN631711
View Documents
MP Biomedicals, Inc
08631711
100μg
Each for $284.50
Add to cart
 
MP8631713
View Documents
MP Biomedicals, Inc
08631713
500μg
Each for $708.50
Add to cart
 
Description

Description

Collagen Type II Monoclonal antibody specifically detects Collagen Type II in Human samples. It is validated for Immunohistochemistry-frozen, Immunohistochemistry-paraffin, Western Blot.
Specifications

Specifications

Collagen Type II
1.0mg/mL
Unconjugated
IgG1
Monoclonal
Store at -20°C. Avoid freeze/thaw cycles.
II-4C11
Purified
Murine
Primary
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit