Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen Type III (N-terminal) Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 227341AP

Catalog No. 50-173-3935


Add to cart

Description

Description

Type I collagen, the fibrillar collagen found in most connective tissues, and the only component of the collagen found in cartilage. Mutations in this gene are associated with osteogenesis imperfecta, Ehlers-Danlos syndrome, and idiopathic osteoporosis. Reciprocal translocations between chromosomes 17 and 22, where this gene and the gene for platelet-derived growth factor beta are located, are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans, resulting from unregulated expression of the growth factor. Two transcripts, resulting from the use of alternate polyadenylation signals, have been identified for this gene.
Specifications

Specifications

Collagen Type III (N-terminal)
0.53 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P02461, P08121, P13941
COL3A1, Collagen alpha 1(III) chain, Collagen Type III, collagen, type III, alpha 1, EDS4A
Rabbit
IgG
150 μL
-20°C
Primary
1281, 12825, 84032
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
COL3A1
Liquid
Col3a1
Collagen Type III (N-terminal) Fusion Protein Ag18658
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit