Learn More
Collagen Type III Rabbit anti-Bovine, Polyclonal, Biologo
Rabbit Polyclonal Antibody
Manufacturer:  Exalpha Biologicals IncSupplier Diversity Partner CO20321
Encompass
 View more versions of this product
Catalog No. 50-210-0727
Add to cart
Description
Description
Collagen Type III Polyclonal antibody specifically detects Collagen Type III in Bovine samples. It is validated for ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Radioimmune Assays, Western Blot
Specifications
Specifications
Collagen Type III
Polyclonal
Unconjugated
Col3a1
Lyophilized
Rabbit
IgG
0.5 mL
-20°C
Primary
510833
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Radioimmune Assays, Western Blot
1 mg/mL
PBS with no preservative; pH
P04258
alpha1 (III) collagen; AW550625; COL 3; COL3; COL3 alpha 1; COL3A1; Col3a-1; Collagen 3; collagen alpha 1 type III; collagen alpha-1(III) chain; collagen type III alpha 1 chain; collagen, fetal; collagen, type III, alpha 1; collagen, type III, alpha 1 (Ehlers-Danlos syndrome type IV, autosomal dominant); collagen, type III, alpha 1 chain, fibril forming; Collagen3; EDS4A; Ehlers-Danlos syndrome type IV, autosomal dominant; FLJ34534; LOC100009177; MGC93704; minisatellite 10w detected by probe MMS10; mKIAA4231; MMS10-W; Ms10w; pro alpha 1 type III collagen; procollagen, type III, alpha 1
Purified collagen type III from bovine skin
Affinity chromatography
RUO
Antibody
Polyclonal
Bovine