Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen Type III Rabbit anti-Chicken, Polyclonal, Biologo Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

$517.28 - $1064.80

Specifications

Antigen Collagen Type III
Concentration 1 mg/mL
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Radioimmune Assays, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-210-0729 Non-distribution product as customer accommodation.
View Documents
Exalpha Biologicals IncSupplier Diversity Partner
CO20331
0.5 mL
Each for $1,064.80
Add to cart
 
50-210-0730 Non-distribution product as customer accommodation.
View Documents
Exalpha Biologicals IncSupplier Diversity Partner
CO2033101
0.1 mL
Each for $517.28
Add to cart
 
Description

Description

Collagen Type III Polyclonal antibody specifically detects Collagen Type III in Chicken samples. It is validated for ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Radioimmune Assays, Western Blot
Specifications

Specifications

Collagen Type III
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Radioimmune Assays, Western Blot
Unconjugated
Rabbit
Chicken
P12105
396340
IgG
Primary
-20°C
Col3a1
1 mg/mL
Polyclonal
Lyophilized
RUO
PBS with no preservative; pH
alpha1 (III) collagen; AW550625; COL 3; COL3; COL3 alpha 1; COL3A1; Col3a-1; Collagen 3; collagen alpha 1 type III; collagen alpha-1(III) chain; collagen type III alpha 1 chain; collagen, fetal; collagen, type III, alpha 1; collagen, type III, alpha 1 (Ehlers-Danlos syndrome type IV, autosomal dominant); collagen, type III, alpha 1 chain, fibril forming; Collagen3; EDS4A; Ehlers-Danlos syndrome type IV, autosomal dominant; FLJ34534; LOC100009177; MGC93704; minisatellite 10w detected by probe MMS10; mKIAA4231; MMS10-W; Ms10w; pro alpha 1 type III collagen; procollagen, type III, alpha 1
Purified collagen type III from chicken skin
Polyclonal
Affinity chromatography
Antibody
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit