Learn More
Advanced Biomatrix Collagen, Human, Type IV, Powder, Non-sterile, 5 mg
Supplier:  Advanced Biomatrix 50165MG
Encompass_Preferred
Type IV Collagen is the primary collagen found in the extracellular basement membranes separating a variety of epithelial and endothelial cells. It is a major component of the dermal-epidermal junction where it is mostly found in the lamina densa. It is a heterotrimeric molecule containing two Alpha 1-like and one Alpha 2-like chains.This Type IV Collagen is isolated from human placenta and is purified using a multi-step process. The product is supplied as a non-sterile powder containing 5 mg of Type IV collagen per vial.Type IV Collagen is typically used as a thin coating on tissue culture surfaces. Specific instructions are found in the Directions for Use. This product is generally used in vitro as a substrate scaffold to enhance cell attachment, adherence and proliferation. Type IV collagen may be used to culture epithelial, endothelial, muscle, nerve and many other cell types.
Catalog No. 50-196-0271
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.