Learn More
Collagen Type IV α2 Chain Mouse anti-Human, Clone: 23IIC3, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma MAB1910I100UG
 View more versions of this product
Catalog No. MAB1910I100
Add to cart
Description
Description
Collagen Type IV α2 Chain Monoclonal antibody specifically detects Collagen Type IV α2 Chain in Human samples. It is validated for Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications
Specifications
Collagen Type IV α2 Chain
23IIC3
Unconjugated
Purified
Purified human amniotic type IV collagen.
Protein G purified
RUO
Primary
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
1.0 mg/mL
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Mouse
IgG1
100 μg
Stable for 1 year at +2°C to +8°C from date of receipt.
Monoclonal