Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen Type IV α2 Chain Mouse anti-Human, Clone: 23IIC3, MilliporeSigma™

Mouse Monoclonal Antibody

$145.80 - $373.00

Specifications

Antigen Collagen Type IV α2 Chain
Clone 23IIC3
Concentration 1.0 mg/mL
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB1910I100
View Documents
MilliporeSigma
MAB1910I100UG
100 μg
Each for $373.00
Add to cart
 
MAB1910I25
View Documents
MilliporeSigma
MAB1910I25UG
25 μg
Each for $145.80
Add to cart
 
Description

Description

Collagen Type IV α2 Chain Monoclonal antibody specifically detects Collagen Type IV α2 Chain in Human samples. It is validated for Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications

Specifications

Collagen Type IV α2 Chain
1.0 mg/mL
Unconjugated
Mouse
Human
Purified human amniotic type IV collagen.
Monoclonal
Protein G purified
23IIC3
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Purified
RUO
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
IgG1
Primary
Stable for 1 year at +2°C to +8°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit