Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen Type IV Rabbit anti-Human, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

Supplier:  Proteintech Group Inc 196741AP150UL

Encompass_Preferred

 View more versions of this product

Catalog No. 50-173-2478


Add to Cart

Description

Description

This antibody is specific to COL4A4.

Collagen IV is a major constituent of the basement membranes along with laminins and enactins. Collagen IV is composed of alpha 1 IV chain and alpha 2 IV chain in 2:1 ratio. Collagen IV can form insoluble fibers with high tensile strength. Antibody to collagen IV is useful in detecting the loss of parts of basement membrane in carcinomas.
Specifications

Specifications

Collagen Type IV
Polyclonal
Unconjugated
COL4A4
CA44, COL4A4, Collagen alpha 4(IV) chain, Collagen Type IV, collagen, type IV, alpha 4
Rabbit
Antigen Affinity Chromatography
RUO
1286
-20°C
Liquid
Immunocytochemistry, Immunofluorescence
0.43 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P53420
COL4A4
Peptide
150 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit