Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen Type VI Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501731332 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-1332 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-1332 Supplier Proteintech Group Inc Supplier No. 170231AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

The extensive family of COL gene products(collagens) iscomposed ofseveral chain types, including fibril-forming interstitialcollagens(typesI, II, III and V) and basement membrane collagens(type IV), each type containing multiple isoforms. Collagens are fibrous, extracellularmatrix proteinswith high tensile strength and are themajorcomponents ofconnective tissue,such astendons and cartilage. Allcollagenscontain a triple helix domain and frequentlyshow lateralself-association in order to formcomplexconnective tissues. Several collagens also play a role in cell adhesion, important formaintaining normal tissue architecture and function.

Specifications

Antigen Collagen Type VI
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene COL6A1
Gene Accession No. P12109, Q04857
Gene Alias COL6A1, collagen 6A1, Collagen alpha 1(VI) chain, collagen, type VI, alpha 1, OPLL
Gene Symbols COL6A1
Host Species Rabbit
Immunogen Collagen Type VI Fusion Protein Ag10288
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12833, 1291, 294337
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.