Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen Type VI Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 148531AP

Catalog No. 50-172-9778


Add to cart

Description

Description

This gene encodes one of the three alpha chains of type VI collagen, a beaded filament collagen found in most connective tissues. The product of this gene contains several domains similar to von Willebrand Factor type A domains. These domains have been shown to bind extracellular matrix proteins, an interaction that explains the importance of this collagen in organizing matrix components. Mutations in this gene are associated with Bethlem myopathy and Ullrich scleroatonic muscular dystrophy. Three transcript variants have been identified for this gene.
Specifications

Specifications

Collagen Type VI
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P12110, Q02788
COL6A2, Collagen alpha 2(VI) chain, Collagen Type VI, collagen, type VI, alpha 2, PP3610
Rabbit
IgG
150 μL
-20°C
Primary
12834, 1292, 361821
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
COL6A2
Liquid
Col6a2
Collagen Type VI Fusion Protein Ag6635
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit