Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen Type VI Rabbit anti-Human, Polyclonal, Biologo Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

$517.28 - $963.16

Specifications

Antigen Collagen Type VI
Concentration 1 mg/mL
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Radioimmune Assays, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-210-0741 Non-distribution product as customer accommodation.
View Documents
Exalpha Biologicals IncSupplier Diversity Partner
CO20611
0.5 mL
Each for $963.16
Add to cart
 
50-210-0742 Non-distribution product as customer accommodation.
View Documents
Exalpha Biologicals IncSupplier Diversity Partner
CO2061101
0.1 mL
Each for $517.28
Add to cart
 
Description

Description

Collagen Type VI Polyclonal antibody specifically detects Collagen Type VI in Human samples. It is validated for ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Radioimmune Assays, Western Blot
Specifications

Specifications

Collagen Type VI
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Radioimmune Assays, Western Blot
Unconjugated
Rabbit
Human
P12109, P12110
1291, 1292
IgG
Primary
-20°C
COL6A2
1 mg/mL
Polyclonal
Lyophilized
RUO
PBS with no preservative; pH
AI747156; alpha 1 (VI) chain (61 AA); BTHLM1; COL6A1; Col6a-1; Col6a2; Col6a-2; Collagen alpha-1(VI) chain; collagen alpha-2(VI) chain; collagen type VI alpha 1; collagen type VI alpha 2; collagen VI, alpha-1 polypeptide; collagen VI, alpha-2 polypeptide; collagen, type VI, alpha 1; collagen, type VI, alpha 2; human mRNA for collagen VI alpha-2 C-terminal globular domain; OPLL; PP3610; procollagen, type VI, alpha 1; procollagen, type VI, alpha 2; RGD1565398; UCHMD1; UCMD1
Purified collagen type VI from human placenta
Polyclonal
Affinity chromatography
Antibody
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit