Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Collagen V Mouse anti-Human, Porcine, Bovine, Canine, Kangaroo, Rabbit, Sheep, Chicken (Negative), Guinea Pig (Negative), Mouse (Negative), Rat (Negative), Clone: 1E2-E4/Col5, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
$176.85 - $436.00
Specifications
Antigen | Collagen V |
---|---|
Clone | 1E2-E4/Col5 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:100-1:2000, ELISA 1:50, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
Description
Collagen V Monoclonal specifically detects Collagen V in Human, Porcine, Bovine, Canine, Kangaroo, Rabbit, Sheep, Chicken (Negative), Guinea Pig (Negative), Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Collagen V | |
1.0 mg/mL | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
Mouse | |
Human, Porcine, Bovine, Canine, Kangaroo, Rabbit, Sheep, Chicken (Negative), Guinea Pig (Negative), Mouse (Negative) | |
1289 | |
Acid-digested pepsin soluble dog type V collagen | |
Primary | |
Store at 4C in the dark. |
1E2-E4/Col5 | |
Western Blot 1:100-1:2000, ELISA 1:50, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
alpha 1 type V collagen, Collagen 5, collagen alpha-1(V) chain, collagen, type V, alpha 1, Collagen-5, EC 2.7.7.6, EC 6.1.1 | |
COL5A1 | |
IgG2a κ | |
Protein A or G purified | |
It has been shown to have no cross reactivity with type I, III and VI collagens by ELISA and immunoblotting. There is no evidence for cross-reactivity with other connective tissue proteins (laminin, fibronectin, elastin). |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title