Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Collagen VI alpha 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP154614
Description
Collagen VI alpha 2 Polyclonal specifically detects Collagen VI alpha 2 in Human samples. It is validated for Western Blot.Specifications
Collagen VI alpha 2 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
collagen alpha-2(VI) chain, collagen VI, alpha-2 polypeptide, collagen, type VI, alpha 2,10PP3610, DKFZp586E1322, FLJ46862 | |
Rabbit | |
109 kDa | |
100 μL | |
Primary | |
Rat: 86%;. | |
Human, Rat, Porcine, Bovine, Canine, Equine | |
Purified |
Western Blot | |
1 mg/ml | |
Western Blot 1.0 ug/ml | |
Q9BUM6 | |
COL6A2 | |
Synthetic peptides corresponding to COL6A2(collagen, type VI, alpha 2) The peptide sequence was selected from the C terminal of COL6A2. Peptide sequence ARRFVEQVARRLTLARRDDDPLNARVALLQFGGPGEQQVAFPLSHNLTAI. | |
Protein A purified | |
RUO | |
1292 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction