Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen VI alpha 2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$116.82 - $343.85

Specifications

Antigen Collagen VI alpha 2
Immunogen Synthetic peptides corresponding to COL6A2(collagen, type VI, alpha 2) The peptide sequence was selected from the C terminal of COL6A2. Peptide sequence ARRFVEQVARRLTLARRDDDPLNARVALLQFGGPGEQQVAFPLSHNLTAI.
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP15461420
View Documents
Novus Biologicals
NBP15461420UL
20ul Each for $116.82
Add to cart
 
NBP154614
View Documents
Novus Biologicals
NBP154614
100ul Each for $343.85
Add to cart
 
Description

Description

Collagen VI alpha 2 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

Collagen VI alpha 2
Rabbit
Polyclonal
Unconjugated
1292
Purified
Western Blot
RUO
Synthetic peptides corresponding to COL6A2(collagen, type VI, alpha 2) The peptide sequence was selected from the C terminal of COL6A2. Peptide sequence ARRFVEQVARRLTLARRDDDPLNARVALLQFGGPGEQQVAFPLSHNLTAI.
Primary
IgG
collagen alpha-2(VI) chain, collagen VI, alpha-2 polypeptide, collagen, type VI, alpha 2,10PP3610, DKFZp586E1322, FLJ46862
PBS and 2% Sucrose with 0.09% Sodium Azide
Protein A purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit