Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen X Mouse anti-Canine, Guinea Pig, Human, Mouse, Porcine, Rat, Clone: X53, eBioscience™ Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  LIFE TECHNOLOGIES 14977195

Encompass

 View more versions of this product

Catalog No. 14-977-195


Add to cart

Description

Description

The monoclonal antibody X53 recognizes human, mouse, porcine, guinea pig, and rat Collagen Type X. X53 does not cross-react with collagen types I, II, III, IV, or VI. Collagen Type X is a homotrimeric short chain collagen. Collagen Type X has limited expression in normal adult tissues, primarily being expressed during bone growth and development. Expression is localized to the peri- and extracellular matrix of hypertrophic chondrocytes during endochondral ossification and also plays a role in cartilage mineralization. Collagen Type X is elevated in many tumor types with expression being localized in the vasculature. Mutations to the gene COL10A1, which encodes human Collagen Type X, is associated with Schmid type metaphyseal chondrodysplasia. Applications Reported: This X53 antibody has been reported for use in microscopy, immunohistochemical staining, ELISA, and immunocytochemistry. Applications Tested: This X53 antibody has been tested by immunohistochemistry on formalin-fixed paraffin embedded human tissue using low pH antigen retrieval and can be used at less than or equal to 10 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

Collagen X Monoclonal antibody specifically detects Collagen X in Canine, Guinea Pig, Human, Mouse, Porcine, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications

Specifications

Collagen X
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q03692, Q05306
Mouse
10 mg
Primary
Canine, Guinea Pig, Human, Mouse, Porcine, Rat
Antibody
IgG1 κ
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
X53
Unconjugated
COL10A1
Col10; Col10a1; Col10a-1; collagen alpha-1(X) chain; collagen type X alpha 1; collagen type X alpha 1 chain; collagen X, alpha-1 polypeptide; collagen, type X, alpha 1; collagen, type X, alpha 1(Schmid metaphyseal chondrodysplasia); pro alpha 1; procollagen, type X, alpha 1; Schmid metaphyseal chondrodysplasia; type X collagen; type X collagen, alpha 1
Affinity chromatography
RUO
100856711, 106026924, 12813, 1300, 25681, 448809
4° C
Liquid
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit