Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Collagen XXV alpha 1 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB300248R
Description
Collagen XXV alpha 1 Polyclonal antibody specifically detects Collagen XXV alpha 1 in Human, Mouse samples. It is validated for Western BlotSpecifications
Collagen XXV alpha 1 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
DyLight 550 | |
Alzheimer disease amyloid-associated protein, AMY, CLAC, CLACP, CLAC-P, Collagen 25, collagen alpha-1(XXV) chain, collagen, type XXV, alpha 1, Collagen-25, collagen-like Alzheimer amyloid plaque component, collagenous Alzheimer amyloid plaque component | |
A synthetic peptide mapping to residues 155-169 [KGEQGDQGPRMVFPK] of the NC2-2 region of human and mouse CLAC-P. | |
0.1 mL | |
Neuroscience | |
84570 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction