Learn More
Abnova™ Collagenase Type 2 (C. histolyticum)
Used for Enzyme
Manufacturer:  Abnova™ P5273
Catalog No. 89-940-153
Add to cart
Description
Description
#REF!
Specifications
Specifications
Native protein purified from Clostridium histolyticum
Lyophilized
Native protein purified from Clostridium histolyticum
Store at 4°C on dry atmosphere. For primary cell isolation, culture and tissue dissociation: Reconstituted in balanced salt solutions (such as Hank's, Earle's and others) at concentrations from 0.05% to 0.5% (w/v). For enzymetic assay: Reconstituted in 0.05 M TES buffer with 0.36 mM calcium chloride, pH 7.5 at a concentration of 1.0 mg/ml.
COLLAGENASE
≥125 units per mg dry weight. One Unit releases one micromole of L-leucine equivalents from collagen in 5 hours at 37°C, pH 7.5.
None
Enzyme Activity
Collagenase Type 2 (C. histolyticum)
100 mg
RUO
Collagenase Assay; Bone, heart, liver, thyroid and salivary primary cell isolation.
Clostridium Species