Learn More
Worthington Biochemical Corporation COLLAGENASE TYPE 4 100MG
SDP
Supplier:  Worthington Biochemical Corporation LS004186
Encompass_Preferred
Collagenase, Type 4,100 MG, Prepared to contain lower tryptic activity levels to limit damage to membrane proteins and receptors but with normal to above normal collagenase activity,Source: Clostridium histolyticum,Store at 2-8degreeC
Catalog No. NC9836075
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.