Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Collagenase, Type I from Clostridium histolyticum, Thermo Scientific Chemicals
Contains mixture of collagenase, clostripain, trypsin, caseinase and other lytic enzymes
$86.10 - $1720.00
Specifications
Concentration | ≥125 U/mg |
---|---|
Components | Contains mixture of collagenase, clostripain, trypsin, caseinase and other lytic enzymes |
Content And Storage | Store desiccated at 4°C |
Source | Clostridium histolyticum |
Product Type | Collagenase |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AAJ13820MC
|
Thermo Scientific
J13820MC |
100 mg |
Each for $86.10
|
|
|||||
AAJ1382003
|
Thermo Scientific
J1382003 |
1 g |
Each for $336.00
|
|
|||||
AAJ1382006
|
Thermo Scientific
J1382006 |
5 g |
Each for $1,720.00
|
|
|||||
Description
- Contains mixture of collagenase, clostripain, trypsin, caseinase and other lytic enzymes
Specifications
≥125 U/mg | |
Store desiccated at 4°C | |
Collagenase |
Contains mixture of collagenase, clostripain, trypsin, caseinase and other lytic enzymes | |
Clostridium histolyticum | |
Lyophilized |
Safety and Handling
EINECSNumber : 232-582-9
Recommended Storage : Keep cold
RUO – Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title