Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Collagenase, Type I from Clostridium histolyticum, Thermo Scientific Chemicals
Contains mixture of collagenase, clostripain, trypsin, caseinase and other lytic enzymes
Supplier: Thermo Scientific J1382006
Description
- Contains mixture of collagenase, clostripain, trypsin, caseinase and other lytic enzymes
Specifications
≥125 U/mg | |
Store desiccated at 4°C | |
5 g | |
Lyophilized |
Contains mixture of collagenase, clostripain, trypsin, caseinase and other lytic enzymes | |
Clostridium histolyticum | |
Collagenase |
Safety and Handling
EINECSNumber : 232-582-9
Recommended Storage : Keep cold
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
RUO – Research Use Only
Spot an opportunity for improvement?
Provide Content Correction