Learn More
Sigma Aldrich Fine Chemicals Biosciences Collagenase from Clostridium histolyticum, 9001-12-1, MFCD00130830, 25 mg DFS Item
Manufacturer:  Sigma Aldrich Fine Chemicals Biosciences C513825MG
Release of physiologically active rat hepatocytes tested, Type IV, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid, Synonym: Clostridiopeptidase A.
Catalog No. NC0217889