Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Collagen V Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP17771750UG
Description
Collagen V Polyclonal specifically detects Collagen V in Human, Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
CollagenV | |
Polyclonal | |
Biotin | |
0.125M Sodium Borate, 0.075M Sodium Chloride, 0.005M EDTA, pH 8.0, 10Mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide | |
COL5A1 | |
Collagen Type V from human and bovine placenta. Uniprot ID P25940 | |
50 μg | |
Primary | |
Negligible cross-reactivity with Type I, II, III, IV or VI collagens. Non-specific cross reaction of anti-collagen antibodies with other human serum proteins or non-collagen extracellular matrix proteins is negligible. | |
Bovine, Human | |
IgG |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
LYOPH | |
Western Blot 1:5000-1:10000, ELISA 1:4000-1:8000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin 1:10-1:500 | |
alpha 1 type V collagen, Collagen 5, collagen alpha-1(V) chain, collagen, type V, alpha 1, Collagen-5, EC 2.7.7.6, EC 6.1.1 | |
Rabbit | |
Affinity Purified | |
Cytoskeleton Markers, Extracellular Matrix | |
1289 | |
Reconstitute with 100μL deionized water. | |
Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction