Learn More
Sklar InstrumentsSupplier Diversity Partner Coller Hemostatic Forceps, Sklar, 7-1/2 in., Curved, Serrated, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 173375
Coller Hemostatic Forceps, Sklar, 7-1/2 in., Curved, Serrated, Stainless Steel. Sklar Coller Hemostatic Forceps, which may also be known as Coller-Crile forceps, are ratcheted, finger ring forceps used for grasping tissue and controlling blood flow in vessels during surgical procedures. They have fully serrated jaws and are available in multiple lengths. This product is curved and 7-1/2 inches long.
Catalog No. 50-143-7413