Learn More
Invitrogen™ Collibri™ 3´ mRNA Library Prep Kit for Illumina™ Systems Non-distribution product as customer accommodation.
Designed for robust construction of cDNA libraries for strand-specific RNA sequencing on Illumina next-generation sequencing (NGS) platforms.
$724.00
Specifications
For Use With (Equipment) Illumina NGS instruments
View More Specs
Products 1
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
A38110024 Non-distribution product as customer accommodation.
View Documents
Invitrogen™
A38110024
24 reactions
Each for $724.00
Add to cart
Description
Description
  • Supports library preparation from polyadenylated RNA in ∼4.5 hours.
  • The protocol generates one fragment per transcript, resulting in accurate gene expression values.
  • Sequences obtained are close to the 3’ ends of the transcripts, so the kit is suitable for varying-quality
  • RNA samples, including formalin-fixed paraffin-embedded (FFPE) samples.
  • The majority of inserts are greater than 75 bp in size, corresponding to final library fragment sizes ≥200 bp.
  • Multiplexing of libraries can be carried out using up to 96 single-indexed primers.
  • For convenience, the Collibri 3’ mRNA Library Prep Kit provides color-coded components for visual tracking of library preparation progress.
  • Inert dyes in the reagents provide visual confirmation of additions and mixing, but do not interfere with enzymatic reactions and do not compromise library prep and sequencing results.
Specifications
Specifications
Illumina NGS instruments