Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Collibri™ Stranded RNA Library Prep Kit for Illumina™ Systems with Human/Mouse/Rat rRNA Depletion Kit Non-distribution product as customer accommodation.

Invitrogen™ Collibri™ Stranded RNA Library Prep Kit for Illumina Systems is designed for robust construction of cDNA libraries for strand-specific RNA sequencing on Illumina next-generation sequencing (NGS) systems.

$2170.00 - $7980.00

Specifications

For Use With (Equipment) Illumina NGS instruments
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
A39003024 Non-distribution product as customer accommodation.
View Documents
Invitrogen™
A39003024
24 preps
Each for $2,170.00
Add to cart
 
A39003096 Non-distribution product as customer accommodation.
View Documents
Invitrogen™
A39003096
96 preps
Each for $7,980.00
Add to cart
 
Description

Description

The kit brings together the superior features of SuperScript IV Reverse Transcriptase, Dynabeads magnetic particles, and Platinum SuperFi™ DNA Polymerase, enabling users to obtain high-quality sequencing-ready libraries.

Stranded RNA Library Prep Kit offers:

  • Short, automation-friendly workflow. Total RNA can be converted to sequencing-ready library within 6.5 hours.
  • Inert dyes in the reagents that make it possible to see the progress of library generation without any negative effect on library quality
  • Superior rRNA removal
  • Uniform transcript coverage and high transcript detection sensitivity
  • High sensitivity of differential gene expression detection
  • >98% strand specificity
  • Efficient detection of non-coding RNA
  • Preservation of 3‘ end sequence information

Designed for fast and convenient library preparation targeting insert sizes of ~150 bp for whole transcriptome libraries starting with sample inputs of 100 to 500 ng of total human, mouse, or rat RNA. Single-tube adapter ligation and reverse transcription reactions, as well as magnetic bead-based rRNA depletion and library purification steps, enable completion of the entire workflow in approximately 6.5 hours. The kit is suitable for various quality RNA samples, including FFPE samples.

For maximum convenience, visual cues of process integrity are included throughout the library prep workflow. Monitor progress of library generation using visual feedback from dyes in critical reagents. The reaction mix changes color in response to the addition of critical components at each step to ensure that library preparation has the highest chance of success. The inert dyes do not interfere with enzymatic reactions and do not compromise sequencing results.

Collibri™ Stranded RNA Library Prep Kit with Human/Mouse/Rat rRNA Depletion Kit includes ribosomal RNA (rRNA) depletion reagents to enable a comprehensive view of the transcriptome through superior removal of human, mouse, and rat rRNA. Full-length Illumina-compatible adaptors are introduced by PCR amplification using Platinum SuperFi™ DNA Polymerase to construct single-indexed libraries compatible with single-read or paired-end sequencing. The kit contains 24 or 96 single-indexed barcoded PCR primers (i7) pre-mixed with a universal i5 primer that enables multiplexing of up to 96 libraries. Optimized cleanup steps efficiently remove residual primers and adapter/primer dimers while preserving high library yields.

A related prioduct, the Collibri™ Library Quantification Kit is recommended for qPCR-based quantifications of libraries before proceeding to sequencing.

Applications:

  • Gene expression studies
  • Alternative splicing analysis
  • Non-coding RNA detection and discovery
  • Identification of alternative polyadenylation sites
  • Gene fusion detection
  • Detection of transcript isoforms
Specifications

Specifications

Illumina NGS instruments
SDS

For Research Use Only. Not for use in diagnostic procedures.

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit