Learn More
Sklar InstrumentsSupplier Diversity Partner Econo Collier Needle Holder, Sklar, 5-1/4 in., Straight, Serrated, Stainless Steel. PK/12. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 21885
Econo Collier Needle Holder, Sklar, 5-1/4 in., Straight, Serrated, Stainless Steel. Econo Collier Needle Holder is a ratcheted, finger ring instrument with fenestrated tips, which some users prefer. The slim profile is an advantage for interproximal areas, and prevents needle rotation or suture slipping. A common field of use is dermatological procedures. Sklar's Econo line of instruments are floor-grade disposables and are sold in bulk quantities.
Catalog No. 50-143-7834