Learn More
Remco™ Color-Coded 8 in. Bench Scraper
Remco™ Color-Coded 8 in. Bench Scraper with one-piece construction that eliminates seams and crevices where bacteria can hide.
Manufacturer:  Remco™ 69602
 View more versions of this product
Catalog No. 17-227-684
Add to cart
Description
Description
  • Ideal solution to your bench top scraping needs
  • Hygienic design has no seams or cracks
  • Helps prevent bacterial growth
Specifications
Specifications
Bench Scraper
Polypropylene
0.9 x 8.3 in.
0.09 lb.
1 Ea.
Medium
Green, Yes
Non-sterile
FDA Compliant