Learn More
Remco™ Color-Coded Coarse Scrub Pad
Remco™ Color-Coded Coarse Scrub Pad is tough on stubborn dirt and grime when cleaning floors.
Manufacturer:  Remco™ 5523
Catalog No. 17-227-474
Add to cart
Description
Description
  • Pairs with the floor tool (5500x) to clean tough messes on poured concrete or tile floors
  • Fits the hand scrubbing tool (5510x) to remove stubborn messes from work benches
  • Is not recommended for use with materials that easily scratch, such as stainless steel
  • Color-coded
Specifications
Specifications
1
Coarse
4.4 in.
No
Medium
Safety
Brown
Polyester Fibers/Abrasive Additives
10 x 4.4 x 0.8 in.
Non-sterile