Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Remco™ Color-Coded Dual Thread Adapter
Remco™ Color-Coded Dual Thread Adapter allows safety shovel blades to be used in conjunction with any European-thread handle.
Manufacturer: Remco™ 6200
Description
- Provides safety tools with a higher degree of flexibility to meet the needs of a variety of safety tasks
- Color-coded
Specifications
White | |
Polypropylene | |
Thread Adapter | |
Non-sterile |
Yes | |
5.4 in. | |
1.25 in. | |
1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title