Learn More
American Educational Products Color-Coded Spring Scales
Clear plastic tube allows students to see the action of the spring.
Manufacturer: American Educational Products 7S40142A
Description
Series of six color-coded tubular spring scales can be used for both force and mass measurements.
- Calibrated in both Newtons and grams with a beveled top for use on an inclined plane
- Incorporate a unique zero adjustment with a clear plastic tube, allowing students to see spring action
Order Info
Available separately or as a set of all six colors
Specifications
250 g, 2.5 N | |
Spring Scale | |
Blue |