Learn More
American Educational Products Color-Coded Spring Scales
SCI_ED
Clear plastic tube allows students to see the action of the spring.
Manufacturer:  American Educational Products 7S40142C
 View more versions of this product
Catalog No. S40142C
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
Series of six color-coded tubular spring scales can be used for both force and mass measurements.
  • Calibrated in both Newtons and grams with a beveled top for use on an inclined plane
  • Incorporate a unique zero adjustment with a clear plastic tube, allowing students to see spring action
Order Info
Available separately or as a set of all six colors
Specifications
Specifications
Spring Scale
1 kg, 10 N
Beige