Learn More
Color-Coded Spring Scales
SCI_ED
Clear plastic tube allows students to see the action of the spring.
Manufacturer:  Si Metric Manufacturing Limited 39600
 View more versions of this product
Catalog No. S02321
Add to cart
Description
Description
Series of eight color-coded tubular spring scales can be used for both force and mass measurements.
  • Calibrated in both Newtons and grams with a beveled top for use on an inclined plane
  • Incorporates a unique zero adjustment
Specifications
Specifications
Blue
4+
0.55 lb. (250 g)
Spring Scale