Learn More
Surgical Design™ Color-Coded Tuning Fork Set With Neurological Hammer
Shop All Surgical Design ProductsDescription
- Complete diagnostic set including multiple color-coded tuning forks and a neurological reflex hammer
- Color-coded silicone handles allow quick identification and improved grip during examinations
- Precision-machined aluminum tuning forks deliver consistent, reliable resonance
- Neurological hammer ideal for reflex testing and comprehensive neuro exams
- Protective zippered case ensures safe storage and easy transport
- Suitable for medical training, neurological exams, audiology testing, and clinical education
Specifications
Specifications
| Product Type | Tuning Fork |
| Certifications/Compliance | ISO/GMP |
| Grade Level | Higher Ed |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.