Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Color Development Reagent, Full Strength, Reagents

Catalog No. 01336517
Click to view available options
Quantity:
1 L
10 L
20 L

Color Development Reagent, Full Strength, Reagent, use for Phosphate analysis. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5, 7647-01-0, 7647-01-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Molybdovanadate reagent
IUPAC Name water
SMILES O

Specifications

Packaging Cubitainer
Quantity 10 L
Specific Gravity 1.141
For Use With (Application) Phosphate Analysis
Physical Form Liquid
Color Green/Yellow
Boiling Point 100°C
For Use With (Equipment) Colorimeter
Solubility Completely miscible
Grade Reagent
Chemical Name or Material Color Development Reagent, Full Strength
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.