Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Color Filter Sets
Explore the science of colors.
$7.50 - $14.00
Specifications
For Use With (Application) | Ideal for use in optical transmission measurements, solar monitoring and exploration, attenuation of laser beams and demonstrations of laser light's monochromacity |
---|
Catalog Number | Mfr. No. | Product Type | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Includes | Price | Quantity & Availability | ||||
S63191
![]() |
Industrial Fiber Optics Inc![]() IFFS1 |
Color Filter Set | Three filters: one red, blue and green |
Each for $7.50
|
|
S63192
![]() |
Industrial Fiber Optics Inc![]() IFFS4 |
Advanced color filter set | 14 Filters: light blue, medium blue, daylight blue, dark blue, golden amber, medium green, light green, green-blue, light red, medium purple, orange, medium yellow, medium red, frost |
Each for $14.00
|
|
Description
Variety of colors individually mounted in 2 x 2 in. (5 x 5cm) 35mm plastic slide holders.
Basic Color Filter Set:
- Three filters: red, blue, green
- 14 filters: light blue, medium blue, daylight blue, dark blue, golden amber, medium green, light green, green-blue, light red, medium purple, orange, medium yellow, medium red, frost
Specifications
Ideal for use in optical transmission measurements, solar monitoring and exploration, attenuation of laser beams and demonstrations of laser light's monochromacity |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title