Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Color Reagent, Solution, For Chloride, APHA, Spectrum™

$58.80 - $206.85

Specifications

CAS 67-56-1, 7782-61-8, 7697-37-2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18-609-887
View Documents
Spectrum Chemical Manufacturing CorporationSupplier Diversity Partner
C330500ML
500 mL Amber Glass Bottle Each for $58.80
Add to cart
 
18-609-888
View Documents
Spectrum Chemical Manufacturing CorporationSupplier Diversity Partner
C3301LT
1 L Amber Glass Bottle Each for $84.00
Add to cart
 
18-609-889
View Documents
Spectrum Chemical Manufacturing CorporationSupplier Diversity Partner
C3304LTGL
4 L Amber Glass Bottle Each for $206.85
Add to cart
 
Specifications

Specifications

67-56-1, 7782-61-8, 7697-37-2
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit